2-Chloro-4-ethoxybenzaldehyde 96%
Lieferant: Apollo Scientific
OR957063-1G
APOSOR957063-1GEA
632
EUR
APOSOR957063-1G
2-Chloro-4-ethoxybenzaldehyde 96%
2-Chloro-4-ethoxybenzaldehyde
Formel:
C₉H₉ClO₂ Molecular Weight: 184,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23132246 CAS-Nummer: 245368-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...