4-Chloro-3,5-dimethylanisole 95%

Lieferant: Apollo Scientific

OR957064-1G
APOSOR957064-1GEA 0 EUR
APOSOR957064-1G
4-Chloro-3,5-dimethylanisole 95%
4-Chloro-3,5-dimethylanisole
Formel: C₉H₁₁ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD07780689
CAS-Nummer: 6981-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR