N-(4-Chlorobenzyl)acetamide 95%
Lieferant: Apollo Scientific
OR957065-1G
APOSOR957065-1GEA
632
EUR
APOSOR957065-1G
N-(4-Chlorobenzyl)acetamide 95%
N-(4-Chlorobenzyl)acetamide
Formel:
C₉H₁₀ClNO Molecular Weight: 183,635 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00757368 CAS-Nummer: 57058-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...