4-Chloro-N-ethylbenzamide 95%

Lieferant: Apollo Scientific

OR957072-1G
APOSOR957072-1GEA 806 EUR
APOSOR957072-1G
4-Chloro-N-ethylbenzamide 95%
4-Chloro-N-ethylbenzamide
Formel: C₉H₁₀ClNO
Molecular Weight: 183.635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00871156
CAS-Nummer: 26930-17-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR