4-(5-Chloropyridin-2-yl)morpholine 95%
Lieferant: Apollo Scientific
OR957086-1G
APOSOR957086-1GEA
538
EUR
APOSOR957086-1G
4-(5-Chloropyridin-2-yl)morpholine 95%
4-(5-Chloropyridin-2-yl)morpholine
Formel:
C₉H₁₁ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD21607482 CAS-Nummer: 59027-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...