3-Chloro-N-propylbenzamide 95%

Lieferant: Apollo Scientific

OR957100-1G
APOSOR957100-1GEA 806 EUR
APOSOR957100-1G
3-Chloro-N-propylbenzamide 95%
3-Chloro-N-propylbenzamide
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01214010
CAS-Nummer: 35306-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR