4-Chloro-N-n-propylbenzamide 95%

Lieferant: Apollo Scientific

OR957101-1G
APOSOR957101-1GEA 806 EUR
APOSOR957101-1G
4-Chloro-N-n-propylbenzamide 95%
4-Chloro-N-n-propylbenzamide
Formel: C₁₀H₁₂ClNO
Molecular Weight: 197,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01212076
CAS-Nummer: 7461-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR