2-(4-Chloro-2,6-dimethylphenoxy)acetonitrile 95%

Lieferant: Apollo Scientific

OR957110-1G
APOSOR957110-1GEA 632 EUR
APOSOR957110-1G
2-(4-Chloro-2,6-dimethylphenoxy)acetonitrile 95%
2-(4-Chloro-2,6-dimethylphenoxy)acetonitrile
Formel: C₁₀H₁₀ClNO
Molecular Weight: 195,646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09944331
CAS-Nummer: 303224-73-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR