2-Chloro-6-naphthol 95%
Lieferant: Apollo Scientific
OR957121-1G
APOSOR957121-1GEA
231
EUR
APOSOR957121-1G
2-Chloro-6-naphthol 95%
2-Chloro-6-naphthol
Formel:
C₁₀H₇ClO Molecular Weight: 178.615 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00127706 CAS-Nummer: 40604-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...