3-Chlor-N,N-dimethylbenzamid 95%

Lieferant: Apollo Scientific

OR957133-1G
APOSOR957133-1GEA 39.3 EUR
APOSOR957133-1G
3-Chlor-N,N-dimethylbenzamid 95%
3-Chlor-N,N-dimethylbenzamid
Formel: C₉H₁₀ClNO
Molecular Weight: 183,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01214035
CAS-Nummer: 24167-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR