1-Chloro-4-methoxynaphthalene 95%

Lieferant: Apollo Scientific

OR957139-1G
APOSOR957139-1GEA 632 EUR
APOSOR957139-1G
1-Chloro-4-methoxynaphthalene 95%
1-Chloro-4-methoxynaphthalene
Formel: C₁₁H₉ClO
Molecular Weight: 192.642 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00053479
CAS-Nummer: 10443-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR