N-(4-Chlorophenylmethyl)tert-butylamine 95%
Lieferant: Apollo Scientific
OR957140-1G
APOSOR957140-1GEA
451
EUR
APOSOR957140-1G
N-(4-Chlorophenylmethyl)tert-butylamine 95%
N-(4-Chlorophenylmethyl)tert-butylamine
Formel:
C₁₁H₁₆ClN Molecular Weight: 197,704 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00600608 CAS-Nummer: 46234-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...