2-Chlor-6-methoxynaphthalin 95%

Lieferant: Apollo Scientific

OR957194-1G
APOSOR957194-1GEA 627 EUR
APOSOR957194-1G
2-Chlor-6-methoxynaphthalin 95%
2-Chloro-6-methoxynaphthalene
Formel: C₁₁H₉ClO
Molecular Weight: 192,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17012373
CAS-Nummer: 67886-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR