3-Benzyloxycarbonylamino-5-nitrobenzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (3-(((Benzyloxy)carbonyl)amino)-5-nitrophenyl)boronic acid, 3-Amino-5-nitrobenzeneboronic acid, CBZ protected

OR9572-1G
APOSOR9572-1GEA 190 EUR
APOSOR9572-1G
3-Benzyloxycarbonylamino-5-nitrobenzeneboronic acid 98%
3-Benzyloxycarbonylamino-5-nitrobenzeneboronic acid
Formel: C₁₄H₁₃BN₂O₆
Molecular Weight: 316,08 g/mol
Schmelzpunkt: 218…220 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235090
CAS-Nummer: 874219-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR