5-Bromopyridine-3-sulphonamide 98%

Lieferant: Apollo Scientific

OR9574-1G
APOSOR9574-1GEA 248 EUR
APOSOR9574-1G
5-Bromopyridine-3-sulphonamide 98%
5-Bromopyridine-3-sulphonamide
Formel: C₅H₅BrN₂O₂S
Molecular Weight: 237,074 g/mol
Schmelzpunkt: 171…173 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235088
CAS-Nummer: 62009-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR