2-Hydroxy-4-methylthiophene 95%

Lieferant: Apollo Scientific

OR957404-1G
APOSOR957404-1GEA 364 EUR
APOSOR957404-1G
2-Hydroxy-4-methylthiophene 95%
2-Hydroxy-4-methylthiophene
Formel: C₅H₆OS
Molecular Weight: 114.166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17078946
CAS-Nummer: 35983-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR