4-Hydroxythiophene-2-carbaldehyde 95%

Lieferant: Apollo Scientific

OR957415-1G
APOSOR957415-1GEA 632 EUR
APOSOR957415-1G
4-Hydroxythiophene-2-carbaldehyde 95%
4-Hydroxythiophene-2-carbaldehyde
Formel: C₅H₄O₂S
Molecular Weight: 128,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26866790
CAS-Nummer: 293742-29-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR