2-Propoxy-3-pyridinol 95%
Lieferant: Apollo Scientific
OR957422-1G
APOSOR957422-1GEA
632
EUR
APOSOR957422-1G
2-Propoxy-3-pyridinol 95%
2-Propoxy-3-pyridinol
Formel:
C₈H₁₁NO₂ Molecular Weight: 153,178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18802323 CAS-Nummer: 1232013-70-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...