2-Methoxypyridin-3-ol 95%

Lieferant: Apollo Scientific

OR957423-5G
APOSOR957423-5GEA 84.1 EUR
APOSOR957423-5G
2-Methoxypyridin-3-ol 95%
2-Methoxypyridin-3-ol
Formel: C₆H₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00457990
CAS-Nummer: 13472-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR