2-Methoxypyrimidin-5-ol 98%

Lieferant: Apollo Scientific

OR957424-1G
APOSOR957424-1GEA 451 EUR
APOSOR957424-1G
2-Methoxypyrimidin-5-ol 98%
2-Methoxypyrimidin-5-ol
Formel: C₅H₆N₂O₂
Molecular Weight: 126,113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696950
CAS-Nummer: 91233-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR