6-(Methylthio)pyridin-3-ol 95%

Lieferant: Apollo Scientific

OR957428-1G
APOSOR957428-1GEA 632 EUR
APOSOR957428-1G
6-(Methylthio)pyridin-3-ol 95%
6-(Methylthio)pyridin-3-ol
Formel: C₆H₇NOS
Molecular Weight: 141,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16999023
CAS-Nummer: 859538-78-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR