1,3-dimethyl-1H-pyrazol-4-ol 95%
Lieferant: Apollo Scientific
OR957431-1G
APOSOR957431-1GEA
574
EUR
APOSOR957431-1G
1,3-dimethyl-1H-pyrazol-4-ol 95%
1,3-dimethyl-1H-pyrazol-4-ol
Formel:
C₅H₈N₂O Molecular Weight: 112,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23110698 CAS-Nummer: 85985-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...