6-Ethoxypyridin-3-ol 95%

Lieferant: Apollo Scientific

OR957434-1G
APOSOR957434-1GEA 632 EUR
APOSOR957434-1G
6-Ethoxypyridin-3-ol 95%
6-Ethoxypyridin-3-ol
Formel: C₇H₉NO₂
Molecular Weight: 139,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18819177
CAS-Nummer: 116178-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR