2-Methoxy-3-methylphenol 95%

Lieferant: Apollo Scientific

OR957438-1G
APOSOR957438-1GEA 661 EUR
APOSOR957438-1G
2-Methoxy-3-methylphenol 95%
2-Methoxy-3-methylphenol
Formel: C₈H₁₀O₂
Molecular Weight: 138,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16998696
CAS-Nummer: 18102-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR