3-Methoxy-4-methylphenol 95%

Lieferant: Apollo Scientific

OR957439-1G
APOSOR957439-1GEA 632 EUR
APOSOR957439-1G
3-Methoxy-4-methylphenol 95%
3-Methoxy-4-methylphenol
Formel: C₈H₁₀O₂
Molecular Weight: 138,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16998731
CAS-Nummer: 19217-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR