2-Methoxy-3-methyl-pyridin-5-ol 95%
Lieferant: Apollo Scientific
OR957442-1G
APOSOR957442-1GEA
364
EUR
APOSOR957442-1G
2-Methoxy-3-methyl-pyridin-5-ol 95%
2-Methoxy-3-methyl-pyridin-5-ol
Formel:
C₇H₉NO₂ Molecular Weight: 139,152 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13705342 CAS-Nummer: 1216253-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...