3-(Methoxymethyl)phenol 95%

Lieferant: Apollo Scientific

OR957443-1G
APOSOR957443-1GEA 538 EUR
APOSOR957443-1G
3-(Methoxymethyl)phenol 95%
3-(Methoxymethyl)phenol
Formel: C₈H₁₀O₂
Molecular Weight: 138,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14680990
CAS-Nummer: 57234-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR