3-(Chloromethyl)phenol 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR957448-1G
OR957448-5G
APOSOR957448-1GEA
484
EUR
APOSOR957448-1G
APOSOR957448-5G
3-(Chloromethyl)phenol 95%
Formel:
C₇H₇ClO Siedepunkt: 105…110 °C (0,1 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD06658536 CAS-Nummer: 60760-06-7 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...