1-Cyclopentyl-1H-pyrazol-5-ol 95%

Lieferant: Apollo Scientific

Synonyme: 1-Cyclopentyl-1h-pyrazol-5-ol

OR957449-1G
APOSOR957449-1GEA 357 EUR
APOSOR957449-1G
1-Cyclopentyl-1H-pyrazol-5-ol 95%
1-Cyclopentyl-1H-pyrazol-5-ol
Formel: C₈H₁₂N₂O
Molecular Weight: 152,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26070663
CAS-Nummer: 1566287-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR