3-Propoxyphenol 95%

Lieferant: Apollo Scientific

Synonyme: 1-Hydroxy-3-propoxybenzene

OR957453-1G
APOSOR957453-1GEA 474 EUR
APOSOR957453-1G
3-Propoxyphenol 95%
3-Propoxyphenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11181933
CAS-Nummer: 6280-96-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR