4-(Propylsulphanyl)phenol 95%

Lieferant: Apollo Scientific

OR957454-1G
APOSOR957454-1GEA 632 EUR
APOSOR957454-1G
4-(Propylsulphanyl)phenol 95%
4-(Propylsulphanyl)phenol
Formel: C₉H₁₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD12780228
CAS-Nummer: 18979-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR