2-Ethoxy-5-methyl-phenol 95%

Lieferant: Apollo Scientific

Synonyme: 2-Ethoxy-5-methylphenol

OR957456-1G
APOSOR957456-1GEA 632 EUR
APOSOR957456-1G
2-Ethoxy-5-methyl-phenol 95%
2-Ethoxy-5-methyl-phenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14534395
CAS-Nummer: 2563-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR