4-Ethoxy-3-methylphenol 95%
Lieferant: Apollo Scientific
OR957457-1G
APOSOR957457-1GEA
632
EUR
APOSOR957457-1G
4-Ethoxy-3-methylphenol 95%
4-Ethoxy-3-methylphenol
Formel:
C₉H₁₂O₂ Molecular Weight: 152,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14535356 CAS-Nummer: 1216257-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...