4-(Methoxymethoxy)phenol 98%
Lieferant: Apollo Scientific
OR957463-1G
APOSOR957463-1GEA
364
EUR
APOSOR957463-1G
4-(Methoxymethoxy)phenol 98%
4-(Methoxymethoxy)phenol
Formel:
C₈H₁₀O₃ Molecular Weight: 154,163 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21195492 CAS-Nummer: 57433-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...