3-(Methoxymethoxy)phenol 95%

Lieferant: Apollo Scientific

OR957464-1G
APOSOR957464-1GEA 0 EUR
APOSOR957464-1G
3-(Methoxymethoxy)phenol 95%
3-(Methoxymethoxy)phenol
Formel: C₈H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD17676551
CAS-Nummer: 18066-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR