2-[(Ethylamino)methyl]phenol 95%
Lieferant: Apollo Scientific
OR957466-1G
OR957466-5G
APOSOR957466-1GEA
400
EUR
APOSOR957466-1G
APOSOR957466-5G
2-[(Ethylamino)methyl]phenol 95%
2-[(Ethylamino)methyl]phenol
Formel:
C₉H₁₃NO Molecular Weight: 151.206 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09929100 CAS-Nummer: 108206-05-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...