2-[(Ethylamino)methyl]phenol 95%

Lieferant: Apollo Scientific

OR957466-1G OR957466-5G
APOSOR957466-1GEA 400 EUR
APOSOR957466-1G APOSOR957466-5G
2-[(Ethylamino)methyl]phenol 95%
2-[(Ethylamino)methyl]phenol
Formel: C₉H₁₃NO
Molecular Weight: 151.206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09929100
CAS-Nummer: 108206-05-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR