2-Chloro-5-(hydroxymethyl)phenol 96%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-3-hydroxybenzyl alcohol
OR957467-1G
APOSOR957467-1GEA
502
EUR
APOSOR957467-1G
2-Chloro-5-(hydroxymethyl)phenol 96%
2-Chloro-5-(hydroxymethyl)phenol
Formel:
C₇H₇ClO₂ Molecular Weight: 158,582 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393156 CAS-Nummer: 1261598-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...