4-(Butylsulphanyl)phenol 95%

Lieferant: Apollo Scientific

OR957473-1G
APOSOR957473-1GEA 632 EUR
APOSOR957473-1G
4-(Butylsulphanyl)phenol 95%
4-(Butylsulphanyl)phenol
Formel: C₁₀H₁₄OS
Molecular Weight: 182,283 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12780229
CAS-Nummer: 1077-25-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR