2-Ethoxy-6-methoxyphenol 95%

Lieferant: Apollo Scientific

OR957475-1G
APOSOR957475-1GEA 0 EUR
APOSOR957475-1G
2-Ethoxy-6-methoxyphenol 95%
2-Ethoxy-6-methoxyphenol
Formel: C₉H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD24721339
CAS-Nummer: 90534-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR