N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine 95%
Lieferant: Apollo Scientific
Synonyme:
6-Chloro-N,N-dimethylpyrazin-2-amine
OR957476-25G
APOSOR957476-25GEA
178
EUR
APOSOR957476-25G
N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine 95%
N-(6-Chloro-2-pyrazinyl)-N,N-dimethylamine
Formel:
C₆H₈ClN₃ Molecular Weight: 157.6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12024889 CAS-Nummer: 61655-72-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...