4-[(Ethoxymethyl)sulphanyl]phenol 95%
Lieferant: Apollo Scientific
OR957477-1G
APOSOR957477-1GEA
632
EUR
APOSOR957477-1G
4-[(Ethoxymethyl)sulphanyl]phenol 95%
4-[(Ethoxymethyl)sulphanyl]phenol
Formel:
C₉H₁₂O₂S Molecular Weight: 184,255 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD21195917 CAS-Nummer: 1468496-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...