2-(Dimethylamino)pyrimidin-5-ol 95%

Lieferant: Apollo Scientific

OR957481-1G
APOSOR957481-1GEA 364 EUR
APOSOR957481-1G
2-(Dimethylamino)pyrimidin-5-ol 95%
2-(Dimethylamino)pyrimidin-5-ol
Formel: C₆H₉N₃O
Molecular Weight: 139,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696945
CAS-Nummer: 72491-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR