5-Methoxy-m-kresol 95%

Lieferant: Apollo Scientific

Synonyme: 3-Hydroxy-5-methylanisol, 1-Hydroxy-3-methoxy-5-methylbenzol, 3-Hydroxy-5-methoxytoluol, 3-Methoxy-5-methylphenol

OR957484-5G
APOSOR957484-5GEA 202 EUR
APOSOR957484-5G
5-Methoxy-m-kresol 95%
5-Methoxy-m-kresol
Formel: C₈H₁₀O₂
Molecular Weight: 138.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059261
CAS-Nummer: 3209-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR