2-[(Propylamino)methyl]phenol 95%

Lieferant: Apollo Scientific

OR957487-1G
APOSOR957487-1GEA 364 EUR
APOSOR957487-1G
2-[(Propylamino)methyl]phenol 95%
2-[(Propylamino)methyl]phenol
Formel: C₁₀H₁₅NO
Molecular Weight: 165,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09951050
CAS-Nummer: 84672-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR