2-(4-Hydroxyphenoxy)acetonitrile 95%

Lieferant: Apollo Scientific

OR957489-1G
APOSOR957489-1GEA 364 EUR
APOSOR957489-1G
2-(4-Hydroxyphenoxy)acetonitrile 95%
2-(4-Hydroxyphenoxy)acetonitrile
Formel: C₈H₇NO₂
Molecular Weight: 149,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11181773
CAS-Nummer: 96562-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR