3-(Propan-2-yloxy)phenol 98%

Lieferant: Apollo Scientific

Synonyme: 3-Isopropoxyphenol

OR957495-1G
APOSOR957495-1GEA 155 EUR
APOSOR957495-1G
3-(Propan-2-yloxy)phenol 98%
3-(Propan-2-yloxy)phenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11103458
CAS-Nummer: 68792-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR