6-Isopropoxypyridin-3-ol 98%

Lieferant: Apollo Scientific

OR957496-1G
APOSOR957496-1GEA 364 EUR
APOSOR957496-1G
6-Isopropoxypyridin-3-ol 98%
6-Isopropoxypyridin-3-ol
Formel: C₈H₁₁NO₂
Molecular Weight: 153,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16997398
CAS-Nummer: 903886-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR