4-(Propan-2-ylsulphanyl)phenol 95%

Lieferant: Apollo Scientific

OR957498-1G
APOSOR957498-1GEA 632 EUR
APOSOR957498-1G
4-(Propan-2-ylsulphanyl)phenol 95%
4-(Propan-2-ylsulphanyl)phenol
Formel: C₉H₁₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD12819457
CAS-Nummer: 70551-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR