(4-Benzamidophenyl)boronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Benzamidobenzeneboronic acid

OR9575-1G
APOSOR9575-1GEA 89.9 EUR
APOSOR9575-1G
(4-Benzamidophenyl)boronsäure 95%
(4-Benzamidophenyl)boronsäure
Formel: C₁₃H₁₂BNO₃
Molecular Weight: 241,05 g/mol
Schmelzpunkt: 194…196 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235089
CAS-Nummer: 397843-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR